missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Tag Monoclonal Antibody (2-2.2.14), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 26183A647
Additional Details : Weight : 0.00010kg
Description
HA Tag Monoclonal antibody specifically detects HA Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.Specifications
HA Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Mouse | |
Protein G | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Immunocytochemistry, Western Blot | |
2-2.2.14 | |
Alexa Fluor 647 | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag | |
50 μL | |
Primary | |
Antibody | |
IgG1 |