Learn More
HA Tag Monoclonal Antibody (2-2.2.14), DyLight™ 800 4X PEG, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 26183D800
Additional Details : Weight : 0.00450kg
Description
26183-D800 has been successfully used for Western blotting. DyLight 800 4X PEG has an excitation/emission of 783/797 nm. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
HA Tag Monoclonal antibody specifically detects HA Tag in Tag samples. It is validated for Western Blot.Specifications
HA Tag | |
Monoclonal | |
1 mg/mL | |
MES with proprietary stabilizer and no preservative | |
Mouse | |
Protein G | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Western Blot | |
2-2.2.14 | |
DyLight 800 4X PEG | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
HA peptide YPYDVPDYA derivitized to ovalbumin. | |
50 μL | |
Primary | |
Antibody | |
IgG1 |