missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Tag Mouse anti-Tag, Clone: 1F5C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66006-2-IG-150UL
Additional Details : Weight : 0.00010kg
Description
HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunoprecipitation of tagged proteins.Specifications
HA Tag | |
Monoclonal | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Mouse | |
Protein G | |
RUO | |
-20°C | |
Liquid | |
Tag |
Immunoprecipitation, Western Blot | |
1F5C6 | |
Unconjugated | |
HA Tag, YPYDVPDYA | |
Peptide | |
150 μL | |
Primary | |
Antibody | |
IgG1 |