missing translation for 'onlineSavingsMsg'
Learn More

HA tag Rabbit anti-Tag, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 51064-2-AP-150UL

Additional Details : Weight : 0.00010kg

Product Code. 16876345

  • £189.00 / 150µL
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunoprecipitation of tagged proteins.
Specifications

Specifications

HA tag
Polyclonal
Unconjugated
HA tag, YPYDVPDYA
Peptide
150 μL
Primary
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.81 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
Antigen Affinity Chromatography
RUO
-20°C
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers