missing translation for 'onlineSavingsMsg'
Learn More

Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Thermo Scientific Chemicals

Product Code. p-4781925
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11307417

Brand: Thermo Scientific Alfa Aesar 012567.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

It is used as an intermediate in chemical research, Pharmaceutical Intermediates,Organic Intermediates. Used in rocket nozzle, especially for re-entry space rocket nose cone.Due to its high hardness and melting point, HfC also widely use in hard alloy additives.Used in ceramic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-85-1
Molecular Formula CHf
Molecular Weight (g/mol) 190.50
MDL Number MFCD00016126
InChI Key NVDNLVYQHRUYJA-UHFFFAOYSA-N
IUPAC Name methanidylidynehafniumylium
SMILES [C-]#[Hf+]

Specifications

Melting Point 3890°C
Color Gray
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.5% (Metals basis excluding Zr)
Solubility Information it is insoluble in water.
Formula Weight 190.54
Odor Odorless
Density 12.2
Chemical Name or Material Hafnium carbide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.