missing translation for 'onlineSavingsMsg'
Learn More
Hafnium crystal bar milled chips, 99.7% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals Product Code.: 11361268

Hafnium crystal bar milled chips, 99.7% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Product Code. 11361268
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11361268

Brand: Thermo Scientific Alfa Aesar 010204.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]

Specifications

Quantity 25 g
Percent Purity 99.7%
Odor Odorless
Form Crystal Bar/Milled Chips
Assay (metals basis excluding Zr), Zr nominal 3%
Chemical Name or Material Hafnium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB11K04 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010204.14,010204.36,010204.22
Lot NumberE31X037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010204.14,010204.36,010204.22
Lot NumberL02M22 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010204.14,010204.36,010204.22
Lot NumberB14Q18 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010204.14,010204.36,010204.22
Lot NumberE18U015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010204.14,010204.36,010204.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hafnium crystal bar milled chips, 99.7% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.