missing translation for 'onlineSavingsMsg'
Learn More

Hafnium dinitrate oxide, 99.9% (metals basis excluding Zr), Zr <0.1%, 10% w/v aq. soln., Thermo Scientific Chemicals

Product Code. p-7052197
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 11349778

Brand: Thermo Scientific Alfa Aesar 044246.AB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00798524

Specifications

Physical Form Liquid
Quantity 10 mL
Linear Formula HfO(NO3)2
UN Number UN3218
Formula Weight 318.5
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 10% w/v aq. soln.
Assay (metals basis excluding Zr), Zr <0.1%
Chemical Name or Material Hafnium dinitrate oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.