missing translation for 'onlineSavingsMsg'
Learn More
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals Product Code.: 11331888

Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals

Product Code. 11331888
500 g
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11331888

Brand: Thermo Scientific Alfa Aesar 011834.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13499-05-3
Molecular Formula Cl4Hf
Molecular Weight (g/mol) 320.29
MDL Number MFCD00011034
InChI Key PDPJQWYGJJBYLF-UHFFFAOYSA-J
Synonym hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis
PubChem CID 37715
IUPAC Name tetrachlorohafnium
SMILES Cl[Hf](Cl)(Cl)Cl

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range >98% (Metals basis excluding Zr)
Formula Weight 320.3
Odor Irritating
Chemical Name or Material Hafnium(IV) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.