missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) chloride, 99%, -80 mesh

Product Code. p-3776923 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10237550

Brand: Thermo Scientific Acros 293250250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13499-05-3 | HfCl4 | 320.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13499-05-3
Molecular Formula HfCl4
Molecular Weight (g/mol) 320.29
MDL Number MFCD00011034
InChI Key PDPJQWYGJJBYLF-UHFFFAOYSA-J
Synonym hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis
PubChem CID 37715
IUPAC Name tetrachlorohafnium
SMILES Cl[Hf](Cl)(Cl)Cl

Specifications

Melting Point 319.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula HfCl4
Merck Index 14, 4588
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol, acetone
Formula Weight 320.29
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Hafnium(IV) chloride, -80 mesh
Show More Show Less
compliance-icons
Product Identifier
  • Hafnium(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.