missing translation for 'onlineSavingsMsg'
Learn More
Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%, Thermo Scientific Chemicals Product Code.: 15417025

Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%, Thermo Scientific Chemicals

Product Code. 15417025
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15417025

Brand: Thermo Scientific Alfa Aesar 045612.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hafnium(IV) chloride is used as a precursor for catalyst involved in the Ziegler-Natta polymerization of propylene. It is an effective Lewis acid for various applications in organic synthesis viz alkylation of ferrocene. Further, it increases the rate and control of 1,3-dipolar cycloadditions. It is also used to prepare hafnium dioxide and hafnium silicate, which is used as high-k-dielectrics in the production of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13499-05-3
Molecular Formula Cl4Hf
Molecular Weight (g/mol) 320.29
MDL Number MFCD00011034
InChI Key PDPJQWYGJJBYLF-UHFFFAOYSA-J
Synonym hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basisShow MoreShow Less
PubChem CID 37715
IUPAC Name tetrachlorohafnium
SMILES Cl[Hf](Cl)(Cl)Cl

Specifications

Melting Point 320° (sublimation)
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in methanol and acetone.
Formula Weight 320.3
Odor Irritating
Sensitivity Moisture sensitive
Chemical Name or Material Hafnium(IV) chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ26K034 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number045612.14,045612.36,045612.22
Lot NumberU15K031 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number045612.14,045612.36,045612.22
Lot NumberW08J011 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number045612.14,045612.36,045612.22
Lot NumberQ23F112 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045612.14,045612.36,045612.22
Lot NumberL30U004 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045612.14,045612.36,045612.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.