missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) fluoride, 99.9% (metals basis excluding Zr), Zr 2-4%, Thermo Scientific™

Product Code. 11317858
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 13709-52-9
MDL Number MFCD00016127

Specifica

Physical Form Sintered Lump
Quantity 25 g
UN Number UN3290
Solubility Information React with water.
Formula Weight 254.48
Percent Purity 99.9%
Sensitivity Moisture sensitive
Assay (metals basis excluding Zr), Zr 2 to 4%
Chemical Name or Material Hafnium(IV) fluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'