missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) fluoride, 99.9% (metals basis excluding Zr), Zr 2-4%, Thermo Scientific™

Product Code. p-7050289
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11307858

Brand: Thermo Scientific Alfa Aesar 036392.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-52-9
MDL Number MFCD00016127

Specifications

Physical Form Sintered Lump
Quantity 5 g
UN Number UN3290
Solubility Information React with water.
Formula Weight 254.48
Percent Purity 99.9%
Sensitivity Moisture sensitive
Assay (metals basis excluding Zr), Zr 2 to 4%
Chemical Name or Material Hafnium(IV) fluoride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.