missing translation for 'onlineSavingsMsg'
Learn More
Hafnium(IV) oxide, 99.995%, (metals basis excluding Zr), Zr typically <0.2%, Thermo Scientific Chemicals Product Code.: 15446955

Hafnium(IV) oxide, 99.995%, (metals basis excluding Zr), Zr typically <0.2%, Thermo Scientific Chemicals

Product Code. 15446955
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15446955

Brand: Thermo Scientific Alfa Aesar 045483.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. It is also used in the preparation of hafnium tetrachloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12055-23-1
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Melting Point 2774°C
Color White
Physical Form Powder
Quantity 5 g
Merck Index 14,4588
Solubility Information Insoluble in water.
Formula Weight 210.49
Percent Purity 99.995%
Odor Odorless
Density 9.68 g/mL
Assay (metals basis excluding Zr), Zr typically <0.2%
Chemical Name or Material Hafnium (IV) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ07J043 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number045483.06,045483.03
Lot NumberZ06H019 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045483.06,045485.BY,045483.03
Lot NumberN10I018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045483.06,045485.BY,045483.03
Lot NumberH26X019 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045483.06,045485.BY,045483.03
Lot NumberR22D082 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number045483.06,045485.BY,045483.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hafnium(IV) oxide, 99.995%, (metals basis excluding Zr), Zr typically <0.2%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.