missing translation for 'onlineSavingsMsg'
Learn More
Hafnium powder, -325 mesh, 99.6% (metals basis excluding Zr), Zr <3.5%, Thermo Scientific Chemicals Product Code.: 11301268

Hafnium powder, -325 mesh, 99.6% (metals basis excluding Zr), Zr <3.5%, Thermo Scientific Chemicals

Product Code. 11301268
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11301268

Brand: Thermo Scientific Alfa Aesar 010201.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]

Specifications

Color Gray
UN Number UN2545
Quantity 5 g
Percent Purity 99.6%
Mesh Size −325 Mesh
Odor Odorless
Form Powder
Assay (metals basis excluding Zr), Zr nominal 2 to 3.5%
Chemical Name or Material Hafnium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24L016 Certificate TypeCertificate of Analysis Date06/03/2025 Catalog Number010201.06,010201.14,010201.22
Lot NumberU22K001 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number010201.06,010201.14,010201.22
Lot NumberS25K027 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number010201.06,010201.14,010201.22
Lot NumberZ22J025 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number010201.06,010201.14,010201.22
Lot NumberU26I020 Certificate TypeCertificate of Analysis Date22/09/2022 Catalog Number010201.06,010201.14,010201.22
compliance-icons
Product Identifier
  • Hafnium powder
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Self-heating substance Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H251-Self-heating: may catch fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P235+P410-Keep cool. Protect from sunlight.
  • P422-Store contents under .
  • P420-Store away from other materials.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hafnium powder, -325 mesh, 99.6% (metals basis excluding Zr), Zr <3.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.