missing translation for 'onlineSavingsMsg'
Learn More

Hafnium silicide, Thermo Scientific Chemicals

£32.15 - £150.75

Chemical Identifiers

CAS 12401-56-8
Molecular Formula HfSi2
Molecular Weight (g/mol) 234.66
MDL Number MFCD00167026
InChI Key TWRSDLOICOIGRH-UHFFFAOYSA-N
Synonym hafnium silicide
PubChem CID 11230071
IUPAC Name hafnium;silicon
SMILES [Si].[Si].[Hf]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387407
View Documents
Thermo Scientific Alfa Aesar
012564.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11377407
View Documents
Thermo Scientific Alfa Aesar
012564.09
10 g
£150.75
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

They have been used as ohmic contacts, Schottky barrier contacts, gate electrodes, local interconnects, and diffusion barriers. FeSi2 has become the remarkable source as high temperature thermoelectric material. It is also used as a powder in ceramic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12401-56-8
234.66
TWRSDLOICOIGRH-UHFFFAOYSA-N
11230071
[Si].[Si].[Hf]
HfSi2
MFCD00167026
hafnium silicide
hafnium;silicon

Specifications

1680°C
Powder
MFCD00167026
it is insoluble in water.
[Si].[Si].[Hf]
234.66
234.66
Hafnium silicide
12401-56-8
HfSi2
hafnium silicide
TWRSDLOICOIGRH-UHFFFAOYSA-N
hafnium;silicon
11230071
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-640-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only