missing translation for 'onlineSavingsMsg'
Learn More

Hafnium telluride, 99.5% (metals basis)

Product Code. 11383618
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11383618

Brand: Thermo Scientific Alfa Aesar 039223.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 39082-23-0 | HfTe2 | 433.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39082-23-0
Molecular Formula HfTe2
Molecular Weight (g/mol) 433.69
MDL Number MFCD00049947
InChI Key BCMXGUYIVFZMCT-UHFFFAOYSA-N
Synonym hafnium telluride, ditellanylidenehafnium
PubChem CID 90471877
IUPAC Name bis(tellanylidene)hafnium
SMILES [Te]=[Hf]=[Te]

Specifications

Physical Form Powder
Quantity 5 g
Formula Weight 433.69
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Hafnium telluride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.