missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium wire, 0.25mm (0.01 in.) dia., 99.97% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 m
2 m
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifica
Specifica
Quantity | 10 m |
Percent Purity | 99.97% |
Odor | Odorless |
Weight | ≈0.64g/m |
Form | Wire |
Assay | (metals basis excluding Zr), Zr nominal 3% |
Chemical Name or Material | Hafnium wire, 0.25mm (0.01 in.) dia. |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T27H017 | Certificate of Analysis | 07/12/2021 | 010200.G2,010200.BW,010200.G6 |
G11T040 | Certificate of Analysis | 24/11/2021 | 010200.G2,010200.BW,010200.G6 |
S26D002 | Certificate of Analysis | 24/11/2021 | 010200.G2,010200.BW,010200.G6 |
61700649 | Certificate of Analysis | 24/11/2021 | 010200.G2,010200.BW,010200.G6 |
61400950 | Certificate of Analysis | 24/11/2021 | 010200.G2,010200.BW,010200.G6 |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Hafnium wire, 0.25mm (0.01 in.) dia., 99.97% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals >
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto