Learn More
3M™ Half Mask Respirator Kits
Brand: 3M™ 7000061547
39.65 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 1.00000kg
Specifications
A12P R | |
Gray | |
Medium | |
Reusable half face mask and set of filters to protect against organic vapors and particulates | |
1/Pk. |
CE Approved | |
Half Mask Respirator Kit | |
Agriculture, Construction, Paint spraying | |
6200 |