Learn More
Medline Scientific™ Half-Shelf Adjuster
Click to view available options
For Use With (Equipment):
BW-10G Heating Bath
Unit Size:
Each
Description
- Manufactured from stainless steel
- With adjustable height support
- Perforated design
- For use with BW-10G General Purpose Heating Baths
Specifications
Specifications
| Description | Perforated |
| For Use With (Equipment) | BW-10G Heating Bath |
| For Use With (Application) | Prevents leaks, guards against rust and contamination |
| Includes | Adjustable height support |
| Material | Stainless Steel |