missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Half Size Food Contact Certified Pallet

Half Size Food Contact Certified Pallet is ideal for equipping preparation area cold rooms.

Brand:  Gilac™ G052260

Product Code. 17948391

  • £178.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Description

Description

  • Food contact certified plastic pallet.
  • Ideal for equipping preparation area cold rooms.
  • Load capacity when stationary: 600 kg
  • Load capacity in motion: 400 kg
  • Storage prohibited on racks: the pads of the pallets are incompatible with the rails of the racks.
Specifications

Specifications

Aero Tote Pallet
800 x 600 x 120 mm
120 mm
800 mm
600 mm
Black
4.7 in.
31.5 in.
23.6 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.