missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Halogen Lamp, 12V/100W, AE31E/BA410E

Product Code. 15435322
Click to view available options
For Use With (Equipment):
AE31E/BA410E Microscopes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15435322

Brand: Motic™ 1101002403781

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Replace worn out microscope bulbs with quality replacement parts. Motic™ Halogen Lamp, 12V/100W, AE31E/BA410E produces light of a higher luminous efficacy and color temperature.

  • For use with AE31E/BA410E Microscopes.
  • Produces light with a higher effective color temperature and higher power efficiency.

Specifications

Type Halogen Bulb
For Use With (Equipment) AE31E/BA410E Microscopes
Voltage 12 V
Wattage 100 W
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.