missing translation for 'onlineSavingsMsg'
Learn More

Halogen Bulb, 20w

Use this light bulb with Max Bino II Compound Microscopes. Fisherbrand™ Halogen Bulbs are ergonomically designed for everyday use.

Brand:  Medline Scientific 2260.6425

Product Code. 12974311

  • ONLINE EXCLUSIVE PRICE
    ÂŁ211.00
    ÂŁ176.94 / Each
    Save ÂŁ34.06
    16% Off
Only null left
In Stock
Edge
Add to basket

Description

Description

  • Halogen bulb, 6V
  • For microscopes: FB69052 or FB69053
Specifications

Specifications

Halogen Bulb
Microscopes: FB69052 or FB69053
6 V
20 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Halogen Bulb, 20w > Halogen Bulb, 20w

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.