Learn More
MP Biomedicals™ Ham's F-10 with L-Glutamine, without Sodium Bicarbonate, Powder
Ham's F-10 powder with L-Glutamine and without Sodium Bicarbonate, used for cell cultures.
Brand:  MP Biomedicals™ 091040120
Product Code. 17308593
  • £154.00 / 10 L
Explore more special offers
Description
Description
Originally, Ham's Nutrient Mixtures were developed for the clonal growth of several Chinese hamster ovary (CHO) clone cells, Hela cells and mouse L-cells. Both F-10 and F-12 mediums are formulated for use with or without serum, depending on the type of cells being cultured.
F-10 has demonstrated satisfactory growth of:
  • Human diploid cells
  • White blood cells for chromosomal analysis
  • Primary rat, rabbit and chicken tissue explants
Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Powder
Cell Culture Medium
Glucose, Glutamine, Phenol Red, Sodium Pyruvate
No Sodium Bicarbonate