missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ham's F-12 Nutrient Mix
GREENER_CHOICE

Ham's F-12 Nutrient Mix

£24.82 - £197.00

Specifications

Cell Line CHO
Concentration 1 X
Form Liquid
Product Type Ham's F-12 Nutrient Mix
With Additives Glutamine, Phenol Red, Sodium Pyruvate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11500586
GREENER_CHOICE
View Documents
Gibco™
21765029
500 mL
£24.82
500mL
Please to purchase this item. Need a web account? Register with us today!
11510586
GREENER_CHOICE
View Documents
Gibco™
21765037
10 x 500 mL
£197.00
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco™ F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ F-12 product made in our Grand Island, New York facility (11765-062). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Gibco™ F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications

Specifications

CHO
Liquid
Glutamine, Phenol Red, Sodium Pyruvate
ISO 13485 standards, FDA
2 to 8°C
1 X
Ham's F-12 Nutrient Mix
Mammalian cell culture
7.1 to 7.5
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Ham's F-12 Nutrient Mix

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.