missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Ham's F-12 Nutrient Mix, GlutaMAX™ Supplement
Ham's F-12 Nutrient Mix, GlutaMAX™ Supplement
Brand: Gibco™ 31765068
Code : 50
Additional Details : Weight : 0.50000kg
Description
Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco™ F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations. Gibco™ F-12 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ F-12 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ F-12 product made in our Grand Island, New York facility (31765-092). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Gibco™ F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
Ham's F-12 Nutrient Mix, 10 x 500mL, 1 X Concentration, Liquid, 7.2 to 7.4 pH, 12 Months Shelf Life, 290 to 320 mOsm/kg Osmolality, With Sodium Pyruvate, GlutaMAX™, Phenol Red, 2 to 8°C Storage, For Mammalian Cell Culture | |
1 X | |
Ham's F-12 Nutrient Mix | |
Mammalian cell culture | |
10 x 500 mL |
2 to 8°C | |
CHO | |
Liquid | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
7.2 to 7.4 |