Learn More
Hamilton™ Modified 7000 Microliter™ Syringes
This series of syringes is ideal for dispensing volumes from 0.05 µL up to 5 µL. The 7000 series is the only zero dead volume syringe. The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle.
£111.00 - £195.00
Specifications
Autoclavable | Autoclavable |
---|---|
Diameter (English) Outer Needle | 0.02 in. |
Diameter (English) Outer | 0.3 in. |
Diameter (Metric) Outer | 7.82 mm |
Length (English) Needle | 2.75 in. |
Product Code | Brand | Volume (Metric) | Needle Gauge | Needle Point Number | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Needle Point Number | Price | Quantity & Availability | |||
10557361
|
Hamilton™
86250 |
0.5 μL | 25 G | 3 |
£148.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10220812
|
Hamilton™
86259 |
0.5 μL | 25 G | 2 |
£195.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10008460
|
Hamilton™
86200 |
1 μL | 22s G | 3 |
£133.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10399322
|
Hamilton™
86211 |
1 μL | 22 G | 2 |
£151.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10323761
|
Hamilton™
88400 |
2 μL | 25 G | 3 |
£141.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10282011
|
Hamilton™
88500 |
2 μL | 23 G | 3 |
£118.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10221921
|
Hamilton™
88011 |
5 μL | 24 G | 2 |
£147.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10401255
|
Hamilton™
88511 |
2 μL | 23 G | 2 |
£158.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10405952
|
Hamilton™
88411 |
2 μL | 25 G | 2 |
£124.00
Each |
Estimated Shipment: 15-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10791232
|
Hamilton™
80100 |
1 μL | 25 G | 3 |
£111.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10332742
|
Hamilton™
88000 |
5 μL | 24 G | 3 |
£174.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- ideal for dispensing volumes from 0.05 µL up to 5 µL
- The 7000 series is the only zero dead volume syringe
- The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Autoclavable | |
0.3 in. | |
2.75 in. | |
239°F | |
50°F | |
Ea. |
0.02 in. | |
7.82 mm | |
General Purpose Syringe | |
115°C | |
10°C | |
Knurled Hub, Removable |