Learn More
Hamilton™ Kel-F Hub Blunt Point Needles (Luer Lock)
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51mm). Custom lengths and point styles are available.
£94.00 - £223.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Stainless Steel |
Length (English) Needle | 2 in. |
Product Type | Kel-F Hub Needle |
Needle Point Style | Blunt |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Needle Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Needle Gauge | Price | Quantity & Availability | |||
13436319
|
Hamilton™
90514 |
0.062 in. | 0.062 in. | 14 G |
£137.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11532325
|
Hamilton™
90521 |
0.02 in. | 0.032 in. | 21 G |
£164.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10007552
|
Hamilton™
90134 |
0.016 in. | 0.028 in. | 22 G |
£223.00
Pack of 6 |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11502325
|
Hamilton™
90516 |
0.046 in. | 0.064 in. | 16 G |
£192.98
Pack of 6 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11512325
|
Hamilton™
90517 |
0.042 in. | 0.057 in. | 17 G |
£97.60
Pack of 6 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11522325
|
Hamilton™
90520 |
0.02 in. | 0.035 in. | 20 G |
£144.00
Pack of 6 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10523515
|
Hamilton™
90523 |
0.013 in. | 0.025 in. | 23 G |
£94.00
Pack of 6 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10136117
|
Hamilton™
90525 |
0.01 in. | 0.01 in. | 25 G |
£94.00
Pack of 6 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10345432
|
Hamilton™
90524 |
0.012 in. | 0.022 in. | 24 G |
£94.00
Pack of 6 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11542325
|
Hamilton™
90528 |
0.007 in. | 0.014 in. | 28 G |
£97.65
Pack of 6 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10790165
|
Hamilton™
90534 |
0.005 in. | 0.028 in. | 22s G |
£122.81
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11552315
|
Hamilton™
90533 |
0.01 in. | 0.018 in. | 26 G |
£94.00
Pack of 6 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11572315
|
Hamilton™
90539 |
0.005 in. | 0.018 in. | 26s G |
£115.00
Pack of 6 |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51mm). Custom lengths and point styles are available.
Kel-F Hub Compatible Syringe Terminations
- Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
- PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
- Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 10 to 31
Six needles per pack.
Specifications
Not Autoclavable | |
2 in. | |
Blunt |
Stainless Steel | |
Kel-F Hub Needle | |
Luer-tip and PTFE Luer-lock Syringes and Connectors |