Learn More
Hamilton™ Large Hub Removable Needles (250μL - 10mL Syringes)
Large Hub Removable (RN) needles can be used with removable needle (RN)-style syringes that are 250μL and larger. Standard needle length is 2 in. (51mm). Custom gauges, lengths, and point styles are available.
£48.05 - £219.00
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Stainless Steel (Hub) |
Length (English) Needle | 2 in. |
Product Type | Replacement Syringe Needle |
For Use With (Equipment) | Removable Syringes (250μL and larger) |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Needle Gauge | Needle Point Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Needle Gauge | Needle Point Style | Price | Quantity & Availability | ||
10103253
|
Hamilton™
7784-04 |
0.01 in. | 0.018 in. | 26 G | Cone |
£179.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10415503
|
Hamilton™
7784-01 |
0.0059 in. | 0.028 in. | 22s G | Cone |
£216.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11570202
|
Hamilton™
7784-03 |
0.0051 in. | 0.018 in. | 26s G | Cone |
£216.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10265092
|
Hamilton™
7779-02 |
0.0051 in. | 0.018 in. | 26s G | Sharp, Bevel, Curved |
£59.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10121812
|
Hamilton™
7779-01 |
0.016 in. | 0.028 in. | 22 G | Sharp, Bevel, Curved |
£128.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10021272
|
Hamilton™
7779-03 |
0.0059 in. | 0.028 in. | 22s G | Sharp, Bevel, Curved |
£62.85
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11507704
|
Hamilton™
7779-04 |
0.01 in. | 0.018 in. | 26 G | Sharp, Bevel, Curved |
£92.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10360901
|
Hamilton™
7780-04 |
0.016 in. | 0.028 in. | 22 G | Blunt |
£107.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10216533
|
Hamilton™
7780-03 |
0.0059 in. | 0.028 in. | 22s G | Blunt |
£48.05
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11517704
|
Hamilton™
7780-01 |
0.0051 in. | 0.018 in. | 26s G | Blunt |
£59.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11527704
|
Hamilton™
7780-02 |
0.01 in. | 0.018 in. | 26 G | Blunt |
£60.75
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10277711
|
Hamilton™
7784-02 |
0.016 in. | 0.028 in. | 22 G | Cone |
£219.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Large Hub Removable Needle Compatible Syringe Terminations
- Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
- Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 26-22
- Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 26-22.
- Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-22
Six needles per pack.
Specifications
Autoclavable | |
2 in. | |
Removable Syringes (250μL and larger) |
Stainless Steel (Hub) | |
Replacement Syringe Needle |