Learn More
Hamilton™ Luer Lock Adapter for Disposable Tips
These adapters enable the connection of a standard 1mL or 200μL disposable pipette tip to the end of a Hamilton Syringe.
Brand:  Hamilton™ 31331
Code : 13
Additional Details : Weight : 0.01000kg
Product Code. 10005640
  • £53.00 / Each
Estimated Shipment: 21-08-2024
to see stock.
Description
Description
  • Two luer tip adapters available: 100μL and 1mL
Specifications
Specifications
CTFE
Luer-tip Adapter
PB600-1 Repeating Dispensers