Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Can be used with TLL syringes and LT or TLL connectors.
£69.50 - £188.00
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Nickel-plated Brass (Hub), Stainless Steel |
Length (English) Needle | 2 in. |
Product Type | Needle |
Needle Point Style | Blunt |
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
11557794
|
Hamilton™
91026 |
26 G |
£104.00
Pack of 6 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11592315
|
Hamilton™
91014 |
14 G |
£135.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11597784
|
Hamilton™
91017 |
17 G |
£113.00
Pack of 6 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11507794
|
Hamilton™
91018 |
18 G |
£111.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11517794
|
Hamilton™
91020 |
20 G |
£118.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15301530
|
Hamilton™
91019 |
19 G |
£69.50
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11527794
|
Hamilton™
91021 |
21 G |
£135.00
Pack of 6 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11537794
|
Hamilton™
91023 |
23 G |
£125.00
Pack of 6 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10422172
|
Hamilton™
91022 |
22 G |
£134.00
Pack of 6 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11547794
|
Hamilton™
91024 |
24 G |
£109.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10775944
|
Hamilton™
91025 |
25 G |
£134.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11572325
|
Hamilton™
91033 |
33 G |
£188.00
Pack of 6 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11587794
|
Hamilton™
91039 |
26s G |
£71.15
Pack of 6 |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10587833
|
Hamilton™
91038 |
22s G |
£71.05
Pack of 6 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Can be used with TLL syringes and LT or TLL connectors.
- Made of 304 Stainless Steel
- Standard needle length is 51mm (2 in.)
- Available in convenient six packs
- Custom needle lengths and point styles are available
- Needles are available from 10 to 33 ga
Specifications
Autoclavable | |
2 in. | |
Blunt |
Nickel-plated Brass (Hub), Stainless Steel | |
Needle |