Learn More
Hamilton™ Microlab 300 Pipetting System Accessories
Accessories for use with Microlab 300 Pipetting System only
Marca: Hamilton™ 66008-01
Code : NEW
Detalles adicionales : Weight : 0.20000kg
Especificaciones
Aluminum | |
Valve Assembly | |
Microlab™ 300 pipetteting system | |
PTFE rotary plug, two 1.5mm threaded ports |