Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
£125.00 - £324.00
Product Code | Brand | Model | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Volume (Metric) | Price | Quantity & Availability | ||||
15142644
|
Hamilton™
59000-05 |
1701 BFP | 10 μL |
£192.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
15122644
|
Hamilton™
59000-15 |
1705 BFP | 50 μL |
£125.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
12638473
|
Hamilton™
59000-20 |
1710 BFP | 100 μL |
£151.00
Each |
Estimated Shipment: 08-11-2024
Log in to see stock. |
||||
15146862
|
Hamilton™
59000-10 |
1702 BFP | 25 μL |
£125.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
15132644
|
Hamilton™
59000-10 |
1702 BFP | 25 μL |
£151.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
12396239
|
Hamilton™
59000-25 |
1725 BFP | 250 μL |
£125.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
13478496
|
Hamilton™
59000-30 |
1750 BFP | 500 μL |
£128.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||||
12528126
|
Hamilton™
59000-35 |
1001 BFP | 1 mL |
£232.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
||||
13346319
|
Hamilton™
59000-40 |
1002 BFP | 2.5 mL |
£216.00
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
11684133
|
Hamilton™
59000-50 |
1010 BFP | 10 mL |
£284.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
15182644
|
Hamilton™
59000-60 |
Bubble free prime | 50 mL |
£324.00
Each |
Estimated Shipment: 11-10-2024
Log in to see stock. |
||||
15192644
|
Hamilton™
59000-55 |
Bubble free prime | 25 mL |
£274.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
12316249
|
Hamilton™
59000-45 |
1005 BFP | 5 mL |
£163.00
Each Qty Price Break Available |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Microlab™ 600 dispensers |