Learn More
Hamilton™ 701N Microliter Syringes
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
Brand:  Hamilton™ 80366
 View more versions of this product
Product Code. 10140221
  • £596.00 / Pack of 6
Edge
Explore more special offers
Description
Description
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Specifications
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
0.019 in.
0.013 cm
0.26 in.
0.66 cm
2 in.
51 mm
137.9 bar
50°C
701 N
10°C
10 μL
Agilent 7670, 7671, and 7672 Automatic Liquid Sampler, Gas Chromatography Autosamplers
26s G
0.005 in.
0.018 in.
0.048 cm
0.047 cm
2 in.
Microliter Autosampler Syringe
122°F
50°F
2
6/Pk.
Sharp, Bevel, Curved, Non-coring
Cemented