missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-X100 Anion Exchange HPLC Columns: Stainless Steel

Polymeric column packing for separation of inorganic and organic anions from 10 to 500 ppm

Brand:  Hamilton™ 79812

Code : NEW

Additional Details : Weight : 0.08000kg

 View more versions of this product

Product Code. 12366509

  • £956.00 / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket

Description

Description

Easy Ion Determination

  • PRP-X100 ion chromatography columns are the easy way to separate inorganic ions.These columns can be used with any existing HPLC or ion chromatograph to easily determine anions in almost any sample matrix including air, water and soil.
Better than Wet Chemical Methods
  • Ion chromatography greatly reduces sample pretreatment and improves the accuracy and precision of results when using wet chemical or colorimetric methods
High Sensitivity
  • Use to separate ions at concentrations from 1 ppm to 1,000 ppm without suppression or elaborate preconcentration techniques
Long Column Life
  • Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), the regeneration protocol will usually return chromatography to its original state.
Analyze Both Difficult and Common Anions
  • Easily separate difficult anions such as cyanide, borate and silicate at high pH (11.5). The polymeric packing is stable from pH 1 to 13, so a single column can be used for the analysis of both common and difficult anions.
Easy Detection and Common Eluents
  • Conductivity detection works by measuring the increased conductance of the eluting sample ions relative to the background conductance

Specifications

Specifications

4.1 mm
Analysis of Fluoride, Carbonate, Chloride, Nitrite, Bromide, Nitrate, Phosphate, Sulfate and Other Ions
5000 psi (345 bar)
5 μm
100 Å
L47
PSDVB/Trimethylammonium
Ion Exchange
Column
1 mL/min
150 mm
pH 1 to 13, 0 to 100% aqueous, organic modifier
1 to 13
5°C to 60°C
PRP-X100
Ion Exchange
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers