missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hammond™ Desiccant, Cole-Parmer™
missing translation for 'orderingAttributeHoverText'
Chemical Name or Material:
Indicating Drierite Desiccant
Non-Indicating Drierite Desiccant
Quantity:
1 lb.
5 lb.
Beschreibung
- Indicating desiccant drying capacity is 10 to 14% of its weight for water and gases
- Non-indicating desiccant drying capacity is 6% of its weight for water and gases
Spezifikation
Spezifikation
Material | Calcium Sulfate |
Quantity | 5 lb. |
Suitability | Used in Desiccators, Drying Columns, or Drying Tubes |
Chemical Name or Material | Non-Indicating Drierite Desiccant |
Sicherheit und Handhabung
Produktidentifikator
- Drierite
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H360F-May damage fertility.
- H412-Harmful to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : cobalt dichloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'