missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hammond™ Drierite™ Desiccant Bags, Cole-Parmer™
Protect materials from corrosion with these desiccant bags. Hammond™ Drierite™ Desiccant Bags reduce the humidity to a 100°F dew point in 10 hours and are dust-tight. Materials packaged with these bags are protected from corrosion for many months.
£130.00 - £193.00
Specifications
Material | Calcium Sulfate |
---|---|
Suitability | These Bags are Dust-Tight and Will Protect For Many Months |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11832692
|
Cole Parmer
07193-04 |
3 g |
£190.00
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11893082
|
Cole Parmer
07193-06 |
6 g |
£173.38
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11803092
|
Cole Parmer
07193-08 |
1 oz. |
£130.00
Pack of 200 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11833092
|
Cole Parmer
07193-14 |
8 oz. |
£193.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nonwoven crepe bags filled with Drierite (anyhydrous calcium sulfate) completely dry the air within a package or container during shipment or storage.- Sturdy and held together by heat seals
- Perform well in elevated temperatures
- Will not release absorbed moisture at less than 350°F
Specifications
Calcium Sulfate | |
These Bags are Dust-Tight and Will Protect For Many Months |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- X750 BAG DRIERITE 3G
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Supplemental information
- MIXTURE LIST-Contains : calcium sulphate