missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hammond™ Stainless Steel Gas Purifier, Cole-Parmer™
Click to view available options
Quantity:
600 mL
Specifications
Specifications
Material | Calcium Sulfate with Cobalt Chloride |
Quantity | 600 mL |
Mesh Size | 40μm |
Packaging | 1 Pack |
Suitability | Low-Cost Gas Purifier Removes Moisture, Oil, and Impurities From Gases Before Analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction