Learn More
CD79b Hamster anti-Mouse, Brilliant Violet 711, Clone: HM79b (also known as HM79-16), BD Optibuild™
Hamster Monoclonal Antibody
Brand: BD Biosciences 740745
341.54 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The HM79b monoclonal antibody specifically recognizes an extracellular epitope of Ig β chain (Ig β or CD79b), a 35-40-kDa transmembrane protein which forms an 80-90-kDa disulfide-linked heterodimer with Ig α chain (Ig α or CD79a, 30-35 kDa). On mature B lymphocytes, the CD79a/CD79b heterodimers are non-covalently associated with surface IgM to form the B-cell receptor complex (BCR). The presence of CD79a/CD79b is necessary for surface expression of the BCR and signal transduction via the BCR in B lymphocytes and pre-B cells. It was recently reported that CD79b may be expressed on the cell surface preceding the appearance of surface IgM during B-lymphocyte development. At this pro-B-cell stage, CD79b participates in signal transduction involved in the regulation of B-cell development. It should be noted that multi-parameter flow cytometric analyses of bone marrow suspensions performed at BD Biosciences Pharmingen have been unable to detect surface staining by HM79b mAb on CD45R/B220+ IgM- cells.

Specifications
CD79b | |
Monoclonal | |
0.2mg/mL | |
Igβ; Igb; Ig-beta; Immunoglobulin-associated beta | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 λ1 |
Flow Cytometry | |
HM79b (also known as HM79-16) | |
Brilliant Violet 711 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
WEHI-123, Mouse B lymphoma | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only