Learn More
Thermo Scientific™ Hand Dispenser Kit
Increase lab flexibility with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems.
Brand: Thermo Scientific™
Code : 03
Additional Details : Weight : 0.20000kg
Description
Increase flexibility in the lab with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems. This kit includes a dispenser with 3 meter cord that connects to the water system tank and a 0.2 micron final filter. It is compatible with Barnstead LabTower™, Pacific™ and Smart2Pure™ systems.
Optional for the following Barnstead water purification systems:- LabTower™ EDI
- LabTower™ RO
- LabTower™ TII
- Smart2Pure™
- Pacific™ TII + tank (tank must have optional pump)
- Pacific™ RO + tank (tank must have optional pump)
Specifications
Hand Dispenser Kit | |
LabTower EDI, LabTower TII, Smart2Pure, Pacific TII + Tank (Tank Must Have Optional Pump), Pacific RO + Tank (Tank Must Have Optional Pump), LabTower RO | |
Hand Dispenser Kit (includes hand dispenser with 3 meter cord, and 0.2 micron final filter) |