missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Hand Operated Vacuum Pumps
Produce up to 25 in Hg vacuum or test for pressure leaks with this vacuum pump. Cole-Parmer™ Hand Operated Vacuum Pumps are sealed and self-lubricated for a long, maintenance-free life.
£145.95 - £178.00
Product Code | Brand | Flow Rate | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Flow Rate | Price | Quantity & Availability | |||||
11785189
|
Cole-Parmer™
79301-01 |
15 mL/stroke |
£145.95
Each |
In Stock
Log in to see stock. |
|||||
11705199
|
Cole-Parmer™
79301-21 |
35 mL/stroke |
£178.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
Description
- Vacuum gauge is graduated in both centimeters and inches of mercury
- Built-in trigger enables you to release vacuum without disconnecting the pump
- Zinc alloy is perfect for rugged applications; PVC is lightweight and easy to handle
Specifications
190.5 x 50.8 x 241.3 mm |