missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ OTAL™ Hand Pump

Product Code. p-8002220 Shop All Bürkle Products
Click to view available options
Flow Rate:
12 L/min.
14 L/min.
20 L/min.
26 L/min.
4 L/min.
Material:
PVDF
Polypropylene
Polypropylene, PVC
Stainless Steel
Unit Size:
Each
This item is not returnable. View return policy
Attention! Make sure to pay attention to the List of Chemical Resistance!

Product Code. 10697331

Brand: Buerkle™ 50051000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Attention! Make sure to pay attention to the List of Chemical Resistance!

Ensure removal of residual liquid with these versatile hand pumps. Buerkle™ OTAL™ Hand Pumps feature a telescopic immersion tube that adapts itself to the container height on insertion, allowing the compact, space-saving hand pump to be used with different containers.

OTAL hand pumps fit most commercial storage drums. The telescopic immersion tube adapts itself to the container height on insertion. This ensures good removal of residual liquid and means that the hand pump can be used with different containers.
  • Particularly compact, space-saving hand pump
  • Good pump capacity
  • With universal bungs made of soft PVC, for container openings of 30–70 mm (1.18-2.76 in.) Ø
OTAL hand pumps with PP delivery tubes:
  • resistant to weak acids, alkalis and detergents.
Pump with PVDF delivery tubes:
  • particularly suitable for acids and alkalis, not for heavily oxidizing acids and alkalis.
  • With its special material properties, PVDF has an even better chemical resistance than PP. Resistant to chlorine bleach.
Stainless steel AISI 304 (1.4301) pump:
  • for liquids that are not easily combustible in Hazard Category AIII (VbF).
  • The OTAL PP or PVDF hand pump should be used for caustic media.
Pump with hose and stopcock:
  • discharge hose with a length of 1.5 m made of PVC and PP/PE stopcock, delivery tube made of PP.
  • The discharge hose is particularly suited to fill distant containers.
TRUSTED_SUSTAINABILITY

Specifications

Applications Commercial Storage
Flow Rate 4 L/min.
Includes 12 mm Dia. Polypropylene Delivery Tube
Weight (Metric) 1 kg
For Use With Storage Drum
Depth (Metric) 45 cm
Pump Tube Length 45 cm
Resistance Properties Alkalis, Detergent, Weak Acid
Type Hand Pump
Drum Opening Diameter 20 to 36 mm
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.