Learn More
Kays Medical Hand Sanitizer Gel
Kays Medical Hand Sanitizer Gel with 70% alcohol is a quick and safe way to sanitize your hands in the workplace.
Brand: Kays Medical BLM3040

Alert:
Use biocides safely. Always read the label and product information before use.
Description
- Refreshing gel formulation
- Quick and highly effective sanitization
- High ethanol content at 70%
- Quick drying with minimal residue
Specifications
Hand Sanitizer | |
Gel | |
Ethanol 70% | |
60 mL |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.