Learn More
Bel-Art™ SP Scienceware™ Handy Stopper/Tubing Size
Easily determines correct size of cork or stopper needed for any flask, bottle
Brand:  Bel-Art™ SP Scienceware™ F13392-0000
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 11392204
  • £14.00 / Each
Estimated Shipment: 13-09-2024
to see stock.
Add to basket
Description
Description
  • Insert thin triangular gauge into vessel and correct stopper size is read at top of neck
  • Two scales provided: one for rubber and vinyl stoppers and the other for corks
  • Back side has circular scale to size tubing outside diameters between 4 and 52mm
Specifications
Specifications
Tubing Sizer
Plastic
Flasks, Bottles