Learn More
SI Analytics™ HandyLab 100 pH Meter HL100/K
Precise, robust and easy to use. Increase measuring speed and accuracy thanks to the HandyLab's AutoRead and CMC function. Supplied in a case with solutions, without electrode.
Brand: SI Analytics™ 285202370
Code : NEW
Additional Details : Weight : 0.40000kg
Description
- Accepts DIN electrodes
- Waterproof IP67
- Reproducible results due to active automatic AutoRead function - eliminates the risk of premature reading
- CMC function to visualize the optimal measuring range
- 1 to 5 point calibration with 22 stored buffer sets
- Data storage (up to 200 data sets) with output on display
- Backlit graphic display with clear text menuM
Specifications
Pocket pH meter HL100/K | |
Carry case and solutions (no electrode) | |
HandyLab 100 |