missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ HandyLab 100 Versatile pH Meter

Product Code. 15354497 Shop All SI Analytics Products
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15354497

Brand: SI Analytics™ 285204520

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SI Analytics™ HandyLab 100 Versatile pH Meter increases the measuring speed and accuracy thanks to the AutoRead and the CMC function.

  • PH-METER Set HandyLab 100 Versatile within suitacse and pHT-combination electrode A7780-NTC30-DIN-N for versatile app
  • AutoRead displays when the measuring value is stable and eliminates the risk of a premature reading of the faulty measuring value
  • CMC (Continuous Measurement Control) visualizes whether the measuring value is still within the calibration limits
  • Having storage capacity for up to 200 data sets, stored data can be viewed
TRUSTED_SUSTAINABILITY

Specifications

Includes 22 preloaded buffer sets
Accuracy (Temperature) ±0.1°C
Accuracy (pH) ±0.005
Range (pH) -2 to 20
Resolution (pH) 0.01
Range (mV) ±1200, ±0.3
Calibration Points 1 to 5
Temperature Range (Metric) -5°C to +105°C
Display Type LCD, B/W Graphic Backlit
Memory Latest calibration
Model HL100Versatile
Power Supply 4 x 1.5 V AA or 4 x 1.2 V NiMH rechargeable battery
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.