missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
Black tips for accurate sample delivery
£179.00 - £715.00
Specifications
Sterility | Non-sterile |
---|---|
Color | Black |
Autoclavable | Autoclavable |
Certifications/Compliance | Non-ANSI |
For Use With (Application) | For accurate sample delivery |
Product Code | Brand | Volume (Metric) | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Format | Price | Quantity & Availability | ||||
11364285
|
Axygen™
TT-1000-CBK-HTR |
1000 μL | Rack |
£179.00
Pack of 1536 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
12521288
|
Axygen™
TT-1000-CBK-HTR-24 |
1000 μL | Rack |
£457.00
Pack of 4608 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
11364295
|
Axygen™
TT-200-CBK-HTR |
200 μL | Rack |
£279.00
Pack of 2304 |
In Stock
Log in to see stock. |
||||
11314295
|
Axygen™
TT-20-CBK-HTR |
20 μL | Rack |
£290.00
Pack of 2304 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
11314305
|
Axygen™
TT-50-CBK-HTR |
50 μL | Rack |
£290.00
Pack of 2304 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
12571318
|
Axygen™
TT-50-CBK-HTR-36 |
50 μL | Rack |
£715.00
Pack of 6912 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
Description
- Polypropylene
- Volume ranging from 20 to 1000μL
- Available with or without aerosol-resistant filters
- Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems
Certifications
Certified DNAse-, RNAse- and Pyrogen-safe
Specifications
Non-sterile | |
Autoclavable | |
For accurate sample delivery | |
Automation, Round | |
Polypropylene |
Black | |
Non-ANSI | |
Rack | |
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head |