Learn More
Corning™ Hank's Balanced Salt Solutions
BIOPHARMA
Salt solutions are shipped at ambient temperature unless otherwise indicated
Brand:  Corning™ 21-020-CV
 View more versions of this product
Product Code. 15353571
  • £103.58 / Pack of 6
Explore more special offers
Description
Description
pH of 10X solutions must be adjusted following dilution to 1X concentration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orange Red
1 X
Hanks' Balanced Salt Solution
7.1 to 7.4
2° to 30°C
Liquid
Calcium, Magnesium
1 x 500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Hank's Balanced Salt Solutions > 1X; Complete Solution; 1 x 500mL