missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Checker™ pH Tester and Replacement Part

For use with virtually any electrode that shares same connector

Brand:  Hanna Instruments™ HI1270

Product Code. 10769741

  • £141.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • pH range from 0.00 to 14.00
  • Provides high accuracy with 0.01 pH resolution at 0° to 50°C
  • Accuracy is ±0.2 pH
  • 1.5V
  • RH max 95%
  • With LCD and manual two point calibration
  • 3000 hours of battery life

Specifications

Specifications

pH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.