Learn More
Hanna Instruments™ pH Electrode Cleaning Solution
Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly. Hanna Instruments™ pH Electrode Cleaning Solution dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
Brand: Hanna Instruments™ HI7061L
Code : A0
Additional Details : Weight : 0.50400kg
Description
General purpose cleaning solution that can be used for a variety of applications.- Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly.
- Dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
Specifications
500 mL | |
Aqueous | |
Removing impurities and residues from ph Electrodes |