Learn More
Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop
Use Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop for general laboratory needs including plating and quick and accurate sample throughput.
Brand: Technical Service Consultants™ TS/4-A
This item is not returnable.
View return policy
Description
- Cadmium-free pigments
- Gamma Irradiation to achieve a SAL of 10-6
- Shelf Life: 3 years
- 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Specifications
Single-use | |
Hard Crystal Polystyrene | |
Clear | |
50/bag |
1 μL | |
Sterile | |
Inoculating Loop | |
Bag |
Product Content Correction
Product Title
Technical Service Consultants™ Hard Crystal Polystyrene Inoculating Loop > Volume: 1μL; Packaging Type: Bag
Spot an opportunity for improvement?Share a Content Correction